siRNA Details
| Gene Symbol: Cd209c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130903.3 | NP_570973.1 | 3 | 1207 | ||
| GenBank mRNA | AF373410.1 | - | 3 | 1207 | ||
| BC120804.1 | - | 3 | 397 | 40129727 | ||
| BC132635.1 | - | 3 | 397 | 40130912 | ||
| BC145088.1 | - | 3 | 397 | 9053636 | ||

Cd209c
Genomic Map