siRNA Details
Gene Symbol: Cd209e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130905.2 | NP_570975.1 | 5, 6 | 514 | ||
GenBank mRNA | AF373412.1 | - | 5, 6 | 514 | ||
AK040917.1 | - | 5, 6 | 536 | |||
BC107188.2 | - | 5, 6 | 514 | 40050810 |
