siRNA Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199198.2 | NP_954668.2 | 3, 4 | 548 | ||
| GenBank mRNA | AK040235.1 | - | 3, 4 | 500 | ||
| AK154045.1 | - | 3, 4 | 512 | |||
| BC013700.1 | - | 3, 4 | 501 | |||
| BC064018.1 | - | 3, 4 | 475 | 3980699 | ||
| BU706947.1 | - | 3, 4 | 289 | |||

Hdac10
Genomic Map