siRNA Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154045.1 | - | 3 | 491 | ||
| BC013700.1 | - | 3 | 480 | |||
| BC064018.1 | - | 3 | 454 | 3980699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdac10| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154045.1 | - | 3 | 491 | ||
| BC013700.1 | - | 3 | 480 | |||
| BC064018.1 | - | 3 | 454 | 3980699 | ||