siRNA Details
| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133239.2 | NP_573502.2 | 4 | 1108 | ||
| GenBank mRNA | AF406641.1 | - | 4 | 1073 | ||
| AY450552.1 | - | 4 | 1073 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crb1| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133239.2 | NP_573502.2 | 4 | 1108 | ||
| GenBank mRNA | AF406641.1 | - | 4 | 1073 | ||
| AY450552.1 | - | 4 | 1073 | |||