siRNA Details
| Gene Symbol: Ahsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133245.1 | NP_573508.1 | 117 | |||
| GenBank mRNA | AF060220.1 | - | 101 | |||
| AF364516.1 | - | 100 | ||||
| AK011084.1 | - | 117 | ||||
| AK028172.1 | - | 117 | ||||
| AK146724.1 | - | 111 | ||||
| BC110627.2 | - | 95 | 40051901 | |||

Ahsp
Genomic Map