siRNA Details
| Gene Symbol: Vgll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133251.1 | NP_573514.1 | 6 | 971 | ||
| GenBank mRNA | AB036708.1 | - | 6 | 971 | ||
| AK146118.1 | - | 5 | 842 | |||
| BC144928.1 | - | 6 | 919 | 9053476 | ||
| BC145949.1 | - | 5 | 903 | 40131252 | ||

Vgll1
Genomic Map