siRNA Details
| Gene Symbol: Inpp5j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172439.3 | NP_766027.2 | 6, 7 | 1848 | ||
| GenBank mRNA | AK034272.1 | - | 6, 7 | 1849 | ||
| AK136908.1 | - | 6, 7 | 1835 | |||
| AK165718.1 | - | 5, 6 | 2376 | |||
| AK170114.1 | - | 6, 7 | 1814 | |||
| AK171029.1 | - | 6, 7 | 1809 | |||
| BC022915.1 | - | 6, 7 | 1801 | |||
| BC046561.1 | - | 5, 6 | 661 | |||
| BC051452.1 | - | 6, 7 | 1817 | |||
| BC131634.1 | - | 6, 7 | 1843 | 40096940 | ||
| BC131635.1 | - | 6, 7 | 1840 | 40096943 | ||

Inpp5j
Genomic Map