siRNA Details
| Gene Symbol: Prima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133364.2 | NP_579942.2 | 3 | 225 | ||
| GenBank mRNA | AK140034.1 | - | 3 | 226 | ||
| AY043275.1 | - | 3 | 226 | |||
| AY225515.1 | - | 3 | 226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prima1| Gene Symbol: Prima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133364.2 | NP_579942.2 | 3 | 225 | ||
| GenBank mRNA | AK140034.1 | - | 3 | 226 | ||
| AY043275.1 | - | 3 | 226 | |||
| AY225515.1 | - | 3 | 226 | |||