siRNA Details
| Gene Symbol: Fut10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012517.5 | NP_001012535.1 | 3, 4 | 770 | ||
| NM_001286422.1 | NP_001273351.1 | 4, 5 | 875 | |||
| NM_001286424.1 | NP_001273353.1 | 3, 4 | 663 | |||
| NM_134161.3 | NP_598922.1 | 3, 4 | 770 | |||
| GenBank mRNA | AJ880009.1 | - | 2, 3 | 383 | ||
| AK147136.1 | - | 4, 5 | 831 | |||
| AK169153.1 | - | 3, 4 | 724 | |||
| AK178457.1 | - | 1, 2 | 251 | |||
| BC030474.1 | - | 3, 4 | 667 | 5368333 | ||
| BC062113.1 | - | 3, 4 | 769 | 6816067 | ||

Fut10
Genomic Map