siRNA Details
| Gene Symbol: Mbnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134163.4 | NP_598924.1 | 5 | 740 | ||
| GenBank mRNA | AB077700.1 | - | 4 | 646 | ||
| AF466292.1 | - | 4 | 646 | |||
| AJ427920.1 | - | 4 | 646 | |||
| AK040522.1 | - | 4 | 494 | |||
| AK050908.1 | - | 4 | 459 | |||
| AK088971.1 | - | 5 | 753 | |||
| AK167342.1 | - | 4 | 459 | |||
| BC057157.1 | - | 5 | 978 | 5348959 | ||

Mbnl3
Genomic Map