siRNA Details
| Gene Symbol: Vmn1r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134176.1 | NP_598937.1 | 1 | 421 | ||
| GenBank mRNA | AK082576.1 | - | 5 | 1071 | ||
| BC132386.1 | - | 1 | 468 | 40130663 | ||
| BC138489.1 | - | 1 | 468 | 8861509 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r4| Gene Symbol: Vmn1r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134176.1 | NP_598937.1 | 1 | 421 | ||
| GenBank mRNA | AK082576.1 | - | 5 | 1071 | ||
| BC132386.1 | - | 1 | 468 | 40130663 | ||
| BC138489.1 | - | 1 | 468 | 8861509 | ||