siRNA Details
| Gene Symbol: Vmn1r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134180.1 | NP_598941.1 | 1 | 784 | ||
| GenBank mRNA | AF394949.1 | - | 940 | |||
| Gene Symbol: Vmn1r18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134181.1 | NP_598942.1 | 1 | 784 | ||
| GenBank mRNA | BC132061.1 | - | 1 | 883 | 40130338 | |
| BC138409.1 | - | 1 | 883 | 8861429 | ||

Vmn1r28
Genomic Map