siRNA Details
| Gene Symbol: Vmn1r19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134182.1 | NP_598943.1 | 1 | 344 | ||
| GenBank mRNA | BC125351.1 | - | 1 | 442 | 40129866 | |
| BC132235.1 | - | 1 | 442 | 40130512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r19| Gene Symbol: Vmn1r19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134182.1 | NP_598943.1 | 1 | 344 | ||
| GenBank mRNA | BC125351.1 | - | 1 | 442 | 40129866 | |
| BC132235.1 | - | 1 | 442 | 40130512 | ||