siRNA Details
| Gene Symbol: Vmn1r9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134185.2 | NP_598946.2 | 1 | 681 | ||
| GenBank mRNA | BC132241.1 | - | 1 | 681 | 40130518 | |
| BC132577.1 | - | 1 | 681 | 40130854 | ||
| Gene Symbol: Vmn1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053231.2 | NP_444461.2 | 1 | 634 | ||

Vmn1r9
Genomic Map