siRNA Details
| Gene Symbol: Vmn1r231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134196.1 | NP_598957.1 | 1 | 239 | ||
| GenBank mRNA | AF394947.1 | - | 308 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r231| Gene Symbol: Vmn1r231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134196.1 | NP_598957.1 | 1 | 239 | ||
| GenBank mRNA | AF394947.1 | - | 308 | |||