siRNA Details
| Gene Symbol: Vmn1r235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134199.3 | NP_598960.3 | 2 | 820 | ||
| GenBank mRNA | AF394953.1 | - | 2 | 821 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r235| Gene Symbol: Vmn1r235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134199.3 | NP_598960.3 | 2 | 820 | ||
| GenBank mRNA | AF394953.1 | - | 2 | 821 | ||