siRNA Details
| Gene Symbol: Vmn1r75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134207.1 | NP_598968.1 | 1 | 246 | ||
| GenBank mRNA | BC115506.1 | - | 1 | 239 | ||
| BC115507.1 | - | 1 | 246 | 40101051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r75| Gene Symbol: Vmn1r75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134207.1 | NP_598968.1 | 1 | 246 | ||
| GenBank mRNA | BC115506.1 | - | 1 | 239 | ||
| BC115507.1 | - | 1 | 246 | 40101051 | ||