siRNA Details
| Gene Symbol: Vmn1r78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134208.2 | NP_598969.2 | 1 | 171 | ||
| GenBank mRNA | BC107194.1 | - | 1 | 171 | 40051037 | |
| BC107195.1 | - | 1 | 171 | 40051039 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r78| Gene Symbol: Vmn1r78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134208.2 | NP_598969.2 | 1 | 171 | ||
| GenBank mRNA | BC107194.1 | - | 1 | 171 | 40051037 | |
| BC107195.1 | - | 1 | 171 | 40051039 | ||