siRNA Details
| Gene Symbol: Vmn1r200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134212.1 | NP_598973.1 | 1 | 451 | ||
| GenBank mRNA | BC131902.1 | - | 1 | 533 | 40130179 | |
| BC131904.1 | - | 1 | 533 | 40130181 | ||
| Gene Symbol: Vmn1r189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145844.1 | NP_665843.1 | 1 | 451 | ||
| GenBank mRNA | BC150715.1 | - | 1 | 550 | 9087626 | |

Vmn1r200
Genomic Map