siRNA Details
| Gene Symbol: Vmn1r199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134213.1 | NP_598974.1 | 1 | 918 | ||
| GenBank mRNA | BC107236.2 | - | 1 | 918 | 40052151 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r199| Gene Symbol: Vmn1r199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134213.1 | NP_598974.1 | 1 | 918 | ||
| GenBank mRNA | BC107236.2 | - | 1 | 918 | 40052151 | |