siRNA Details
| Gene Symbol: Vmn1r214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134214.1 | NP_598975.1 | 1 | 352 | ||
| GenBank mRNA | BC145835.1 | - | 1 | 396 | 40131138 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r214| Gene Symbol: Vmn1r214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134214.1 | NP_598975.1 | 1 | 352 | ||
| GenBank mRNA | BC145835.1 | - | 1 | 396 | 40131138 | |