siRNA Details
Gene Symbol: Vmn1r205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134217.1 | NP_598978.1 | 1 | 219 | ||
GenBank mRNA | BC132016.1 | - | 1 | 289 | 40130293 | |
BC138103.1 | - | 1 | 289 | 8861122 |
