siRNA Details
| Gene Symbol: Vmn1r218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134222.1 | NP_598983.1 | 1 | 127 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r218| Gene Symbol: Vmn1r218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134222.1 | NP_598983.1 | 1 | 127 | ||