siRNA Details
| Gene Symbol: Vmn1r195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134223.2 | NP_598984.2 | 1 | 103 | ||
| GenBank mRNA | BC116946.1 | - | 1 | 103 | 40126265 | |
| BC119333.1 | - | 1 | 103 | 8734082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r195| Gene Symbol: Vmn1r195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134223.2 | NP_598984.2 | 1 | 103 | ||
| GenBank mRNA | BC116946.1 | - | 1 | 103 | 40126265 | |
| BC119333.1 | - | 1 | 103 | 8734082 | ||