siRNA Details
| Gene Symbol: Vmn1r193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134225.1 | NP_598986.1 | 1 | 879 | ||
| Gene Symbol: Vmn1r204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045544.1 | NP_001039009.1 | 1 | 828 | ||

Vmn1r193
Genomic Map