siRNA Details
| Gene Symbol: Vmn1r70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134228.1 | NP_598989.1 | 1 | 544 | ||
| GenBank mRNA | AF394955.1 | - | 1 | 544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r70| Gene Symbol: Vmn1r70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134228.1 | NP_598989.1 | 1 | 544 | ||
| GenBank mRNA | AF394955.1 | - | 1 | 544 | ||