siRNA Details
| Gene Symbol: Vmn1r66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134230.3 | NP_598991.2 | 1 | 876 | ||
| GenBank mRNA | AF394952.1 | - | 1 | 875 | ||
| BC109341.1 | - | 1 | 729 | 40052743 | ||
| BC109342.1 | - | 1 | 729 | 40052744 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r66| Gene Symbol: Vmn1r66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134230.3 | NP_598991.2 | 1 | 876 | ||
| GenBank mRNA | AF394952.1 | - | 1 | 875 | ||
| BC109341.1 | - | 1 | 729 | 40052743 | ||
| BC109342.1 | - | 1 | 729 | 40052744 | ||