siRNA Details
| Gene Symbol: Vmn1r185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134231.1 | NP_598992.1 | 1 | 385 | ||
| GenBank mRNA | BC150720.1 | - | 1 | 471 | 9087631 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r185| Gene Symbol: Vmn1r185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134231.1 | NP_598992.1 | 1 | 385 | ||
| GenBank mRNA | BC150720.1 | - | 1 | 471 | 9087631 | |