siRNA Details
| Gene Symbol: Vmn1r203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134236.1 | NP_598997.1 | 1 | 833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r203| Gene Symbol: Vmn1r203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134236.1 | NP_598997.1 | 1 | 833 | ||