siRNA Details
| Gene Symbol: Vmn1r211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134243.1 | NP_599004.1 | 1 | 437 | ||
| GenBank mRNA | BC138182.1 | - | 1 | 485 | 8861201 | |
| BC138183.1 | - | 1 | 485 | 8861202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r211| Gene Symbol: Vmn1r211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134243.1 | NP_599004.1 | 1 | 437 | ||
| GenBank mRNA | BC138182.1 | - | 1 | 485 | 8861201 | |
| BC138183.1 | - | 1 | 485 | 8861202 | ||