siRNA Details
| Gene Symbol: Vmn1r216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134245.1 | NP_599006.1 | 1 | 817 | ||
| GenBank mRNA | BC145650.1 | - | 1 | 895 | 40130953 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r216| Gene Symbol: Vmn1r216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134245.1 | NP_599006.1 | 1 | 817 | ||
| GenBank mRNA | BC145650.1 | - | 1 | 895 | 40130953 | |