siRNA Details
| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134250.2 | NP_599011.2 | 4 | 579 | ||
| GenBank mRNA | AF399831.1 | - | 4 | 525 | ||
| AF450241.1 | - | 4 | 575 | |||
| AK148474.1 | - | 4 | 580 | |||
| AK170775.1 | - | 5 | 399 | |||
| AY553334.1 | - | 4 | 572 | |||
| BC106851.2 | - | 4 | 537 | 40046221 | ||
| BC106852.2 | - | 4 | 537 | 40046222 | ||

Havcr2
Genomic Map