siRNA Details
| Gene Symbol: Slc22a27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134256.1 | NP_599017.1 | 8 | 1228 | ||
| GenBank mRNA | AB056442.1 | - | 8 | 1228 | ||
| AB056443.1 | - | 6 | 877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a27| Gene Symbol: Slc22a27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134256.1 | NP_599017.1 | 8 | 1228 | ||
| GenBank mRNA | AB056442.1 | - | 8 | 1228 | ||
| AB056443.1 | - | 6 | 877 | |||