siRNA Details
| Gene Symbol: Slc26a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134420.4 | NP_599252.2 | 2508 | |||
| GenBank mRNA | AF248494.1 | - | 1854 | |||
| AK029238.1 | - | 3430 | ||||
| AK046993.1 | - | 3768 | ||||
| AK053088.1 | - | 2159 | ||||
| AK163670.1 | - | 2509 | ||||
| AK163671.1 | - | 2509 | ||||
| AY049076.1 | - | 1768 | ||||
| CN692543.1 | - | 328 | ||||

Slc26a6
Genomic Map