siRNA Details
| Gene Symbol: Il17rd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134437.3 | NP_602319.1 | 5 | 581 | ||
| GenBank mRNA | AF459444.1 | - | 5 | 500 | ||
| AF494209.1 | - | 1 | 290 | |||
| AF494210.1 | - | 5 | 641 | |||
| AK133233.1 | - | 7 | 744 | |||
| AK139904.1 | - | 5 | 506 | |||
| BC066804.1 | - | 4 | 991 | |||
| BC138629.1 | - | 5 | 536 | 8861649 | ||

Il17rd
Genomic Map