siRNA Details
| Gene Symbol: Il17rd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134437.3 | NP_602319.1 | 11 | 1243 | ||
| GenBank mRNA | AF424804.1 | - | 7 | 672 | ||
| AF459444.1 | - | 11 | 1162 | |||
| AF494209.1 | - | 7 | 952 | |||
| AF494210.1 | - | 11 | 1303 | |||
| AK133233.1 | - | 13 | 1406 | |||
| AK139904.1 | - | 11 | 1169 | |||
| BC138629.1 | - | 11 | 1198 | 8861649 | ||

Il17rd
Genomic Map