siRNA Details
| Gene Symbol: Creld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133930.1 | NP_598691.1 | 9 | 1364 | ||
| GenBank mRNA | AB009020.1 | - | 9 | 1369 | ||
| AK050160.1 | - | 2 | 145 | |||
| AK050455.1 | - | 2 | 145 | |||
| AK077726.1 | - | 9 | 1509 | |||
| AK155777.1 | - | 9 | 1374 | |||
| BC010804.1 | - | 9 | 1364 | 4239786 | ||
| BC023058.1 | - | 2 | 242 | |||
| BC023893.1 | - | 9 | 1375 | 5310111 | ||
| BC025932.1 | - | 9 | 1270 | 5102571 | ||
| BC029065.1 | - | 9 | 1400 | 4935296 | ||

Creld1
Genomic Map