siRNA Details
| Gene Symbol: Dicer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148948.2 | NP_683750.2 | 13 | 2305 | ||
| GenBank mRNA | AB081470.1 | - | 12 | 2103 | ||
| AF430845.1 | - | 13 | 2313 | |||
| AK129241.1 | - | 7 | 864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dicer1| Gene Symbol: Dicer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148948.2 | NP_683750.2 | 13 | 2305 | ||
| GenBank mRNA | AB081470.1 | - | 12 | 2103 | ||
| AF430845.1 | - | 13 | 2313 | |||
| AK129241.1 | - | 7 | 864 | |||