siRNA Details
| Gene Symbol: Mvd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138656.2 | NP_619597.2 | 1 | 117 | ||
| GenBank mRNA | AJ309922.1 | - | 1 | 74 | ||
| AK089354.1 | - | 1 | 119 | |||
| AK134803.1 | - | 1 | 106 | |||
| AK153439.1 | - | 1 | 92 | |||
| AK154883.1 | - | 1 | 119 | |||
| AK170007.1 | - | 1 | 54 | |||
| AK170773.1 | - | 1 | 113 | |||
| BC008526.1 | - | 1 | 85 | 3495424 | ||

Mvd
Genomic Map