siRNA Details
| Gene Symbol: Nlgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163387.1 | NP_001156859.1 | 6 | 2307 | ||
| NM_138666.3 | NP_619607.2 | 8 | 3334 | |||
| GenBank mRNA | AK122433.1 | - | 8 | 3335 | ||
| AK138209.1 | - | 3 | 1168 | |||
| BC098461.1 | - | 6 | 2307 | 30099449 | ||

Nlgn1
Genomic Map