siRNA Details
| Gene Symbol: Flna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290421.1 | NP_001277350.1 | 40 | 6689 | ||
| NM_010227.3 | NP_034357.2 | 39 | 6667 | |||
| GenBank mRNA | AF353671.1 | - | 3 | 368 | ||
| AK131179.1 | - | 27 | 4508 | |||
| BC004061.1 | - | 2 | 188 | |||
| BC038478.1 | - | 40 | 6564 | |||
| BC039208.1 | - | 19 | 2879 | |||
| BC151024.1 | - | 39 | 6658 | 9087937 | ||
| BQ950333.1 | - | 3 | 259 | |||

Flna
Genomic Map