siRNA Details
| Gene Symbol: Flna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290421.1 | NP_001277350.1 | 45 | 7486 | ||
| NM_010227.3 | NP_034357.2 | 44 | 7464 | |||
| GenBank mRNA | AK131179.1 | - | 32 | 5305 | ||
| BC004061.1 | - | 7 | 985 | |||
| BC038478.1 | - | 45 | 7361 | |||
| BC039208.1 | - | 24 | 3676 | |||
| BC054432.1 | - | 1 | 64 | |||
| BC099506.1 | - | 2 | 236 | |||
| BC151024.1 | - | 44 | 7455 | 9087937 | ||

Flna
Genomic Map