siRNA Details
| Gene Symbol: Stab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138672.2 | NP_619613.2 | 30 | 3361 | ||
| GenBank mRNA | AB052959.1 | - | 30 | 3282 | ||
| AF290914.2 | - | 30 | 3292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stab1| Gene Symbol: Stab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138672.2 | NP_619613.2 | 30 | 3361 | ||
| GenBank mRNA | AB052959.1 | - | 30 | 3282 | ||
| AF290914.2 | - | 30 | 3292 | |||