siRNA Details
| Gene Symbol: Pkhd1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138674.2 | NP_619615.2 | 24 | 2792 | ||
| GenBank mRNA | AB055648.1 | - | 24 | 2792 | ||
| AY219182.1 | - | 24 | 2792 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkhd1l1| Gene Symbol: Pkhd1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138674.2 | NP_619615.2 | 24 | 2792 | ||
| GenBank mRNA | AB055648.1 | - | 24 | 2792 | ||
| AY219182.1 | - | 24 | 2792 | |||