siRNA Details
| Gene Symbol: Shkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138676.2 | NP_619617.2 | 6 | 432 | ||
| GenBank mRNA | AF246218.1 | - | 6 | 438 | ||
| AK047434.1 | - | 4 | 505 | |||
| AK154779.1 | - | 6 | 421 | |||
| AK160641.1 | - | 5 | 504 | |||
| AK166328.1 | - | 5 | 377 | |||
| AK177754.1 | - | 1 | 18 | |||
| BC061477.1 | - | 6 | 385 | 6336438 | ||

Shkbp1
Genomic Map