siRNA Details
| Gene Symbol: Shkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138676.2 | NP_619617.2 | 14, 15 | 1541 | ||
| GenBank mRNA | AF246218.1 | - | 14, 15 | 1547 | ||
| AK047434.1 | - | 12, 13 | 1672 | |||
| AK047826.1 | - | 2, 3 | 340 | |||
| AK154779.1 | - | 14, 15 | 1529 | |||
| AK160641.1 | - | 13, 14 | 1613 | |||
| AK182187.1 | - | 1, 2 | 72 | |||
| AK186318.1 | - | 1, 2 | 112 | |||
| AK187576.1 | - | 1, 2 | 72 | |||
| BC061477.1 | - | 14, 15 | 1494 | 6336438 | ||

Shkbp1
Genomic Map