siRNA Details
| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138680.2 | NP_619621.2 | 8, 9 | 1210 | ||
| GenBank mRNA | AF318301.1 | - | 8, 9 | 1135 | ||
| AK078853.1 | - | 54 | ||||
| BC002314.1 | - | 3, 4 | 145 | |||
| BC020058.1 | - | 9, 10 | 935 | 4009362 | ||
| BC040793.1 | - | 7, 8 | 702 | |||
| BC054449.1 | - | 7, 8 | 1153 | |||
| BC056354.1 | - | 8, 9 | 1208 | 6811460 | ||
| BC056970.1 | - | 8, 9 | 1208 | 6811460 | ||

Luc7l2
Genomic Map