siRNA Details
| Gene Symbol: Lrrc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138682.2 | NP_619623.2 | 1 | 1829 | ||
| GenBank mRNA | AF300458.1 | - | 1 | 1786 | ||
| AK046781.1 | - | 1 | 1036 | |||
| BC012209.1 | - | 1 | 1830 | |||
| BC117833.1 | - | 1 | 1770 | 40092947 | ||
| BC117834.1 | - | 1 | 1770 | 40092948 | ||
| DQ177325.1 | - | 1 | 1829 | |||

Lrrc4
Genomic Map