siRNA Details
| Gene Symbol: Prom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138750.2 | NP_620089.2 | 14 | 1802 | ||
| NM_178047.4 | NP_835148.2 | 14 | 1802 | |||
| GenBank mRNA | AF269062.1 | - | 14 | 1852 | ||
| AK029851.1 | - | 14 | 1784 | |||
| AK137749.1 | - | 14 | 1780 | |||
| BC039212.1 | - | 14 | 1770 | 4188940 | ||

Prom2
Genomic Map